2-Hydroxy-5-(trifluoromethyl)pyridine ≥98.0%

Supplier: TCI America

Synonyms: 5-(Trifluoromethyl)pyridin-2-ol

T2539-25G T2539-5G
TCT2539-25GEA 208.56 CAD
TCT2539-25G TCT2539-5G
2-Hydroxy-5-(trifluoromethyl)pyridine ≥98.0%
2-Hydroxy-5-(trifluoromethyl)pyridine

CAS Number: 33252-63-0
MDL Number: MFCD00042315
Molecular Formula: C6H4F3NO
Molecular Weight: 163.10
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 149


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 5-(Trifluoromethyl)-2-pyridinone, 2-Hydroxy-5-(trifluoromethyl)pyridine

Formula: C₆H₄F₃NO
MW: 163.1
Melting Pt: 149 °C
MDL Number: MFCD00042315
CAS Number: 33252-63-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db160f994e3b923afa3d6af49fb4092b0144b6aa&vid=db160f994e3b923afa3d6af49fb4092b0144b6aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR