Tween® 85 (Polysorbate) for biochemical research

Supplier: TCI America

Synonyms: Polyoxyethylenesorbitan Trioleate

T2534-25G T2534-5G
TCT2534-25GEA 56.25 CAD
TCT2534-25G TCT2534-5G
Tween® 85 (Polysorbate) for biochemical research
Tween® 85 (Polysorbate)

[for Biochemical Research]
CAS Number: 9005-70-3
MDL Number: MFCD01779641
Form: Clear Liquid
Color: Pale Yellow
Boiling point (°C): 100
Melting point (°C): -20
Flash Point (°C): 315
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: Polyoxyethylene Sorbitan Trioleate

Boiling Pt: 100 °C
Melting Pt: –20 °C
Density: 1.03 (20 °C)
Flash Pt: 315 °C
MDL Number: MFCD01779641
CAS Number: 9005-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR