2,2'-Bithiophene-5-boronic acid pinacol ester ≥98.0%

Supplier: TCI America

T2518-1G T2518-5G T2518-25G
TCT2518-1GEA 71.1 CAD
TCT2518-1G TCT2518-5G TCT2518-25G
2,2'-Bithiophene-5-boronic acid pinacol ester ≥98.0%
2,2'-Bithiophene-5-boronic acid pinacol ester

CAS Number: 479719-88-5
MDL Number: MFCD04039974
Molecular Formula: C14H17BO2S2
Molecular Weight: 292.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Blue


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives, Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2-(2,2'-Bithiophen-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2,2'-Bithiophene-5-boronic Acid Pinacol Ester

Formula: C₁₄H₁₇BO₂S₂
MDL Number: MFCD04039974
CAS Number: 479719-88-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e796a91cd4eb4f7b63a2788e8e957087aedaba45&vid=e796a91cd4eb4f7b63a2788e8e957087aedaba45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR