Thieno[3,2-b]thiophene ≥98.0%
Supplier: TCI America
T2513-1G
T2513-5G
TCT2513-1GEA
158
CAD
TCT2513-1G
TCT2513-5G
Thieno[3,2-b]thiophene ≥98.0%
Thieno[3,2-b]thiophene
CAS Number: 251-41-2
MDL Number: MFCD00179479
Molecular Formula: C6H4S2
Molecular Weight: 140.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Boiling point (°C): 71
Melting point (°C): 56
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...