N-(Triphenylmethyl)-DL-serine methyl ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N-(Triphenylmethyl)-DL-serine Methyl Ester, N-Trityl-DL-serine Methyl Ester, Trt-DL-Ser-OMe, Methyl 3-hydroxy-2-(tritylamino)propanoate

T2500-5G T2500-25G
TCT2500-5GEA 45.82 CAD
TCT2500-5G TCT2500-25G
N-(Triphenylmethyl)-DL-serine methyl ester ≥98.0% (by GC, titration analysis)
N-(Triphenylmethyl)-DL-serine methyl ester

CAS Number: 13515-76-9
Molecular Formula: C23H23NO3
Molecular Weight: 361.44
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 146


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: N-Trityl-DL-serine Methyl Ester, Trt-DL-Ser-OMe

Formula: C₂₃H₂₃NO₃
MW: 361.44 g/mol
Melting Pt: 146 °C
MDL Number: MFCD00583950
CAS Number: 13515-76-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=974b70ecf8ae195cc5633e772b209b122e09bd61&vid=974b70ecf8ae195cc5633e772b209b122e09bd61&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR