1H,1H-Tridecafluoroheptyl iodide ≥95.0%

Supplier: TCI America

Synonyms: 1H,1H-Perfluoro-1-iodoheptane, 1H,1H-Tridecafluoro-1-iodoheptane, 1H,1H-Perfluoroheptyl Iodide

T2482-1G T2482-5G
TCT2482-1GEA 154.84 CAD
TCT2482-1G TCT2482-5G
1H,1H-Tridecafluoroheptyl iodide ≥95.0%
1H,1H-Tridecafluoroheptyl iodide

CAS Number: 212563-43-4
MDL Number: MFCD00233026
Molecular Formula: C7H2F13I
Molecular Weight: 459.98
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 2.02


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1H,1H-Perfluoro-1-iodoheptane, 1H,1H-Tridecafluoro-1-iodoheptane, 1H,1H-Perfluoroheptyl Iodide

Formula: C₇H₂F₁₃I
Density: 2.02 (20 °C)
MDL Number: MFCD00233026
CAS Number: 212563-43-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=80b3ffe136146ae222bde879bfa18180ad9b30c0&vid=80b3ffe136146ae222bde879bfa18180ad9b30c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR