4-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde ≥97.0%
Supplier: TCI America
T2481-1G
T2481-5G
TCT2481-1GEA
67.94
CAD
TCT2481-1G
TCT2481-5G
4-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde ≥97.0%
4-(1,1,2,2-Tetrafluoroethoxy)benzaldehyde
CAS Number: 35295-36-4
MDL Number: MFCD00039577
Molecular Formula: C9H6F4O2
Molecular Weight: 222.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 103
Melting point (°C): 9
Flash Point (°C): 110
Specific Gravity (20/20): 1.39
Formula:
C₉H₆F₄O₂ Boiling Pt: 103 °C Melting Pt: 9 °C Density: 1.39 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00039577 CAS Number: 35295-36-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...