4,9,11-Trioxatetracyclo[5.3.1.0(2,6).0(8,10)]undecan-3-one ≥98.0%

Supplier: TCI America

T2471-1G T2471-5G
TCT2471-1GEA 183.28 CAD
TCT2471-1G TCT2471-5G
4,9,11-Trioxatetracyclo[5.3.1.0(2,6).0(8,10)]undecan-3-one ≥98.0%
4,9,11-Trioxatetracyclo[5.3.1.0(2,6).0(8,10)]undecan-3-one

CAS Number: 632339-23-2
Molecular Formula: C8H8O4
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 181


Other Product Names / Synonyms: 2,6-Epoxy-2,2a,5,5a,6,6a-hexahydrooxireno[f]isobenzofuran-3(1aH)-one

Formula: C₈H₈O₄
Melting Pt: 181 °C
CAS Number: 632339-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a7158fe737da21966b30111c9cd36c74e5d52a0&vid=9a7158fe737da21966b30111c9cd36c74e5d52a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR