7-(Trifluoromethyl)isatin ≥98.0%

Supplier: TCI America

T2457-5G T2457-1G
TCT2457-5GEA 102.7 CAD
TCT2457-5G TCT2457-1G
7-(Trifluoromethyl)isatin ≥98.0%
7-(Trifluoromethyl)isatin

CAS Number: 391-12-8
MDL Number: MFCD01248408
Molecular Formula: C9H4F3NO2
Molecular Weight: 215.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: Yellow
Melting point (°C): 193


Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction), Fluorinated Building Blocks
Other Product Names / Synonyms: 7-(Trifluoromethyl)indole-2,3-dione, 7-(Trifluoromethyl)-2,3-indolinedione

Formula: C₉H₄F₃NO₂
MW: 215.1300 g/mol
Melting Pt: 193 °C
MDL Number: MFCD01248408
CAS Number: 391-12-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5cc7277ce4aedc119c3b7a4377e9eeec4485f0d5&vid=5cc7277ce4aedc119c3b7a4377e9eeec4485f0d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR