L-Tyrosine-tert-butyl ester ≥97.0% (by HPLC, titration analysis)
Supplier: TCI America
T2446-25G
T2446-5G
TCT2446-25GEA
364.98
CAD
TCT2446-25G
TCT2446-5G
L-Tyrosine-tert-butyl ester ≥97.0% (by HPLC, titration analysis)
L-Tyrosine-tert-butyl ester
CAS Number: 16874-12-7
MDL Number: MFCD00042644
Molecular Formula: C13H19NO3
Molecular Weight: 237.30
Purity/Analysis Method: >97.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 140
Specific rotation [a]20/D: 25 deg (C=2, EtOH)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Tyr-OtBu
Formula:
C₁₃H₁₉NO₃ Melting Pt: 140 °C |
MDL Number:
MFCD00042644 CAS Number: 16874-12-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...