(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: H-D-Trp-OMe·HCl, D-Tryptophan methyl ester hydrochloride, H-D-Trp-OMe.HCl

T2442-25G T2442-5G
TCT2442-25GEA 159.58 CAD
TCT2442-25G TCT2442-5G
(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride ≥98.0% (by total nitrogen basis)
(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride

CAS Number: 14907-27-8
MDL Number: MFCD00038992
Molecular Formula: C12H14N2O2
Molecular Weight: 254.71
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 205
Specific rotation [a]20/D: -19 deg (C=5, MeOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-D-Trp-OMe.HCl

Formula: C₁₂H₁₄N₂O₂·HCl
MW: 254.7 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00038992
CAS Number: 14907-27-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa5237f7c180d9aff233fbac1f9ba728266f3ca7&vid=aa5237f7c180d9aff233fbac1f9ba728266f3ca7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR