L-Threonine-tert-butyl ester hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: (2S,3R)-tert-Butyl 2-amino-3-hydroxybutanoate hydrochloride, H-Thr-OtBu.HCl

T2435-1G T2435-5G
TCT2435-1GEA 53.72 CAD
TCT2435-1G TCT2435-5G
L-Threonine-tert-butyl ester hydrochloride ≥98.0%
L-Threonine-tert-butyl ester hydrochloride

CAS Number: 69320-90-7
MDL Number: MFCD00672361
Molecular Formula: C8H17NO3
Molecular Weight: 211.69
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Color: White
Specific rotation [a]20/D: -12 deg (C=1, MeOH)


Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: H-Thr-OtBu.HCl

Formula: C₈H₁₇NO₃
MW: 211.69 g/mol
MDL Number: MFCD00672361
CAS Number: 69320-90-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=183b9d250f87565b4619465efd22f1d52143bc46&vid=183b9d250f87565b4619465efd22f1d52143bc46&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR