Trimethyl-1,2,3-propanetricarboxylate ≥97.0%

Supplier: TCI America

Synonyms: 3-(Methoxycarbonyl)pentanedioic Acid Dimethyl Ester, Dimethyl 3-(Methoxycarbonyl)pentanedioate, 1,2,3-Propanetricarboxylic Acid Trimethyl Ester

T2429-5G T2429-1G
TCT2429-5GEA 158 CAD
TCT2429-5G TCT2429-1G
Trimethyl-1,2,3-propanetricarboxylate ≥97.0%
Trimethyl-1,2,3-propanetricarboxylate

CAS Number: 6138-26-7
MDL Number: MFCD00040552
Molecular Formula: C9H14O6
Molecular Weight: 218.21
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 150
Specific Gravity (20/20): 1.18


Other Product Names / Synonyms: 1,2,3-Propanetricarboxylic Acid Trimethyl Ester, Dimethyl 3-(Methoxycarbonyl)pentanedioate, 3-(Methoxycarbonyl)pentanedioic Acid Dimethyl Ester

Formula: C₉H₁₄O₆
MW: 218.21 g/mol
Boiling Pt: 150 °C
Density: 1.18 (20 °C)
MDL Number: MFCD00040552
CAS Number: 6138-26-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa24ea12ce1e395d27b0ab2f57a61ab40323d400&vid=fa24ea12ce1e395d27b0ab2f57a61ab40323d400&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR