trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
trans-4-(Trifluoromethyl)cyclohexane-carboxylicacid
T2424-1G
T2424-5G
TCT2424-1GEA
75.84
CAD
TCT2424-1G
TCT2424-5G
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
trans-4-(Trifluoromethyl)cyclohexanecarboxylic acid
CAS Number: 133261-33-3
Molecular Formula: C8H11F3O2
Molecular Weight: 196.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 154
Formula:
C₈H₁₁F₃O₂ MW: 196.17 g/mol Melting Pt: 154 °C |
MDL Number:
MFCD09037363 CAS Number: 133261-33-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...