cis-4-(Trifluoromethyl)cyclohexanecarboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: cis-4-(Trifluoromethyl)cyclohexane-1-carboxylic acid

T2423-1G T2423-5G
TCT2423-1GEA 191.18 CAD
TCT2423-1G TCT2423-5G
cis-4-(Trifluoromethyl)cyclohexanecarboxylic acid ≥98.0%
cis-4-(Trifluoromethyl)cyclohexanecarboxylic acid

CAS Number: 1202578-27-5
Molecular Formula: C8H11F3O2
Molecular Weight: 196.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 99

Formula: C₈H₁₁F₃O₂
MW: 196.17 g/mol
Melting Pt: 99 °C
MDL Number: MFCD09037361
CAS Number: 1202578-27-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d36e38c32db05adee0f449d327d2e748ceabe547&vid=d36e38c32db05adee0f449d327d2e748ceabe547&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR