1,2,4-Triazole-3-carboxamide ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 2H-1,2,4-Triazole-3-carboxamide

T2402-25G T2402-5G
TCT2402-25GEA 453.46 CAD
TCT2402-25G TCT2402-5G
1,2,4-Triazole-3-carboxamide ≥97.0% (by HPLC, total nitrogen)
1,2,4-Triazole-3-carboxamide

CAS Number: 3641-08-5
MDL Number: MFCD03990481
Molecular Formula: C3H4N4O
Molecular Weight: 112.09
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Color: White
Melting point (°C): 315

Formula: C₃H₄N₄O
MW: 112.09 g/mol
Melting Pt: 315 °C
MDL Number: MFCD03990481
CAS Number: 3641-08-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d1d9bd9df97c70492ec69207e8130d5386f6059&vid=0d1d9bd9df97c70492ec69207e8130d5386f6059&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR