N-(Trimethylsilyl)bis(trifluoromethanesulfonyl)imide ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
TMS Triflimide, [Bis(trifluoromethanesulfonyl)amino]trimethylsilane, [Bis(trifluoromethanesulphonyl)amino]trimethylsilane
CAS Number: 82113-66-4
Molecular Formula: C5H9F6NO4S2Si
Molecular Weight: 353.33
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 115
Storage Temperature: <0°C
Product Categories: Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: [Bis(trifluoromethanesulfonyl)amino]trimethylsilane, TMS Triflimide
Formula:
C₅H₉F₆NO₄S₂Si MW: 353.33 g/mol Boiling Pt: 115 °C |
MDL Number:
MFCD10566930 CAS Number: 82113-66-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...