2,3-Thiophenedicarboxaldehyde ≥98.0%

Supplier: TCI America

T2388-5G T2388-1G
TCT2388-5GEA 529.3 CAD
TCT2388-5G TCT2388-1G
2,3-Thiophenedicarboxaldehyde ≥98.0%
2,3-Thiophenedicarboxaldehyde

CAS Number: 932-41-2
MDL Number: MFCD00209616
Molecular Formula: C6H4O2S
Molecular Weight: 140.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Boiling point (°C): 134
Melting point (°C): 79

Formula: C₆H₄O₂S
Boiling Pt: 134 °C
Melting Pt: 79 °C
MDL Number: MFCD00209616
CAS Number: 932-41-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00da317965c5f1f0caa7f75c72293f16b559be10&vid=00da317965c5f1f0caa7f75c72293f16b559be10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR