2,4,5-Trifluoro-4-hydroxybenzoic acid ≥98.0%

Supplier: TCI America

T2380-25G T2380-5G
TCT2380-25GEA 191.18 CAD
TCT2380-25G TCT2380-5G
2,4,5-Trifluoro-4-hydroxybenzoic acid ≥98.0%
2,4,5-Trifluoro-4-hydroxybenzoic acid

CAS Number: 116751-24-7
MDL Number: MFCD00800378
Molecular Formula: C7H3F3O3
Molecular Weight: 192.09
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 146

Formula: C₇H₃F₃O₃
MW: 192.09
Melting Pt: 146 °C
MDL Number: MFCD00800378
CAS Number: 116751-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b4127110f25670cb379a41ac72f5fdcd9dd37c86&vid=b4127110f25670cb379a41ac72f5fdcd9dd37c86&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR