4'-Methylbiphenyl-2-carboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: 4'-Methyl-2-biphenylcarboxylic acid, 4'-Methyl-[1,1'-biphenyl]-2-carboxylic acid

T2377-5G T2377-25G
TCT2377-5GEA 28.44 CAD
TCT2377-5G TCT2377-25G
4'-Methylbiphenyl-2-carboxylic acid ≥98.0%
4'-Methylbiphenyl-2-carboxylic acid

CAS Number: 7148-03-0
MDL Number: MFCD00045826
Molecular Formula: C14H12O2
Molecular Weight: 212.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 150


Other Product Names / Synonyms: 2-(4-Methylphenyl)benzoic Acid, 4'-Methylbiphenyl-2-carboxylic Acid

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Melting Pt: 150 °C
MDL Number: MFCD00045826
CAS Number: 7148-03-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fad13865c899faf3677343dc5ec427fd785a11aa&vid=fad13865c899faf3677343dc5ec427fd785a11aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR