1,5,6,7-Tetrahydro-2-methyl-4H-indol-4-one ≥98.0%

Supplier: TCI America

T2376-25G T2376-5G
TCT2376-25GEA 56.88 CAD
TCT2376-25G TCT2376-5G
1,5,6,7-Tetrahydro-2-methyl-4H-indol-4-one ≥98.0%
1,5,6,7-Tetrahydro-2-methyl-4H-indol-4-one

CAS Number: 35308-68-0
MDL Number: MFCD04972500
Molecular Formula: C9H11NO
Molecular Weight: 149.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 204


Other Product Names / Synonyms: 2-Methyl-4-oxo-4,5,6,7-tetrahydroindole

Formula: C₉H₁₁NO
Melting Pt: 204 °C
MDL Number: MFCD04972500
CAS Number: 35308-68-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=090010dfc5c164483349c5c5af6b71e356382b91&vid=090010dfc5c164483349c5c5af6b71e356382b91&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR