2,2,3,3-Tetramethylcyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

T2350-25G T2350-5G
TCT2350-25GEA 97.49 CAD
TCT2350-25G TCT2350-5G
2,2,3,3-Tetramethylcyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
2,2,3,3-Tetramethylcyclopropanecarboxylic acid

CAS Number: 15641-58-4
MDL Number: MFCD00011538
Molecular Formula: C8H14O2
Molecular Weight: 142.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 120

Formula: C₈H₁₄O₂
Melting Pt: 120 °C
MDL Number: MFCD00011538
CAS Number: 15641-58-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d219d72f7e2d1c78da538eb2fdaf1a723d8aa14&vid=7d219d72f7e2d1c78da538eb2fdaf1a723d8aa14&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR