3-Pyridineboronic acid pinacol ester ≥97.0%

Supplier: TCI America

Synonyms: Pyridine-3-boronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

T2345-5G T2345-1G T2345-25G
TCT2345-5GEA 131.14 CAD
TCT2345-5G TCT2345-1G TCT2345-25G
3-Pyridineboronic acid pinacol ester ≥97.0%
3-Pyridineboronic acid pinacol ester

CAS Number: 329214-79-1
MDL Number: MFCD03084758
Molecular Formula: C11H16BNO2
Molecular Weight: 205.06
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 104


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 2-(3-Pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Pyridylboronic Acid Pinacol Ester

Formula: C₁₁H₁₆BNO₂
MW: 205.06 g/mol
Melting Pt: 104 °C
MDL Number: MFCD03084758
CAS Number: 329214-79-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dd0f04cd9b164c6054998e5f27102b10f9e63ca&vid=1dd0f04cd9b164c6054998e5f27102b10f9e63ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR