3,3,4,4-Tetrafluoropyrrolidine hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

T2342-1G T2342-100MG
TCT2342-1GEA 438.72 CAD
TCT2342-1G TCT2342-100MG TCT2342-5G
3,3,4,4-Tetrafluoropyrrolidine hydrochloride ≥98.0% (by total nitrogen and titration analysis)
3,3,4,4-Tetrafluoropyrrolidine hydrochloride

CAS Number: 1810-13-5
Molecular Formula: C4H5F4N
Molecular Weight: 179.54
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Color: White

Formula: C₄H₅F₄N
MDL Number: MFCD06200835
CAS Number: 1810-13-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6f01f6152e2e28242f5a724bb89c68d52453c39&vid=e6f01f6152e2e28242f5a724bb89c68d52453c39&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR