4,4',4''-Triaminotriphenylamine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Tris(4-aminophenyl)amine
T2332-1G
T2332-5G
TCT2332-1GEA
160.37
CAD
TCT2332-1G
TCT2332-5G
4,4',4''-Triaminotriphenylamine ≥98.0% (by HPLC, titration analysis)
4,4',4''-Triaminotriphenylamine
CAS Number: 5981-09-9
MDL Number: MFCD00778301
Molecular Formula: C18H18N4
Molecular Weight: 290.37
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 230
Formula:
C₁₈H₁₈N₄ MW: 290.38 g/mol Melting Pt: 230 °C |
MDL Number:
MFCD00778301 CAS Number: 5981-09-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...