Trimethyl[3-(triethoxysilyl)propyl]ammonium chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

T2246-25G T2246-5G
TCT2246-25GEA 1128.12 CAD
TCT2246-25G TCT2246-5G
Trimethyl[3-(triethoxysilyl)propyl]ammonium chloride ≥98.0% (by titrimetric analysis)
Trimethyl[3-(triethoxysilyl)propyl]ammonium chloride

CAS Number: 84901-27-9
Molecular Formula: C12H30ClNO3Si
Molecular Weight: 299.91
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White

Formula: C₁₂H₃₀ClNO₃Si
CAS Number: 84901-27-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2d1f28ab3854b4afd7c491322d9a1e0eaffa416&vid=f2d1f28ab3854b4afd7c491322d9a1e0eaffa416&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR