2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: (2R,3R,4S,5R,6R)-2-(Acetoxymethyl)-6-isothiocyanatotetrahydro-2H-pyran-3,4,5-triyl triacetate, GITC

A5514-1G A5514-100MG
TCA5514-001GEA 990.66 CAD
TCA5514-001G TCA5514-100MG
2,3,4,6-Tetra-O-acetyl-β-D-glucopyranosyl isothiocyanate ≥98.0% (by HPLC, total nitrogen)
2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranosyl isothiocyanate

[for HPLC Labeling]
CAS Number: 14152-97-7
MDL Number: MFCD00043085
Molecular Formula: C15H19NO9S
Molecular Weight: 389.38
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 116


Product Categories: Bioscience, Glucose, Glycosides, O-Substituted Sugars, Analytical Science, e.e. Determination (HPLC Labeling Reagents), UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: GITC

Formula: C₁₅H₁₉NO₉S
MW: 389.38 g/mol
Melting Pt: 116 °C
MDL Number: MFCD00043085
CAS Number: 14152-97-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dc5511bc38e425b0e6740367a4cbed01654c80b&vid=9dc5511bc38e425b0e6740367a4cbed01654c80b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR