Rubrene ≥99.0%, purified by sublimation

Supplier: TCI America

Synonyms: 5,6,11,12-Tetraphenylnaphthacene, 5,6,11,12-Tetraphenyltetracene

T2233-1G T2233-250MG
TCT2233-1GEA 420.28 CAD
TCT2233-1G TCT2233-250MG
Rubrene ≥99.0%, purified by sublimation
Rubrene

CAS Number: 517-51-1
MDL Number: MFCD00003703
Molecular Formula: C42H28
Molecular Weight: 532.69
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Color: Pale Yellow
Melting point (°C): 331


Product Categories: Materials Chemistry, Organic Transistor Materials, Organic Light-Emitting Diode (OLED) Materials, Nanocarbon Unit Structures (Nanocarbon Materials), Highly Purified Reagents
Other Product Names / Synonyms: Rubrene

Formula: C₄₂H₂₈
MW: 532.68 g/mol
Melting Pt: 331 °C
MDL Number: MFCD00003703
CAS Number: 517-51-1
EINECS: 208-242-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a327e93b62cb28dc86f35003e48c32a69762fa46&vid=eb4ac8be840a321473efd01ca18122ddf9ac092a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR