9-(p-Tolyl)carbazole ≥98.0%

Supplier: TCI America

T2227-5G T2227-1G
TCT2227-5GEA 241.27 CAD
TCT2227-5G TCT2227-1G
9-(p-Tolyl)carbazole ≥98.0%
9-(p-Tolyl)carbazole

CAS Number: 19264-73-4
Molecular Formula: C19H15N
Molecular Weight: 257.34
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: Very Pale Yellow
Melting point (°C): 107

Formula: C₁₉H₁₅N
Melting Pt: 107 °C
MDL Number: MFCD14708175
CAS Number: 19264-73-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=951bd5d2cd4a634879f17432390b5f9edde82b3d&vid=951bd5d2cd4a634879f17432390b5f9edde82b3d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR