5,10,15,20-Tetra(4-pyridyl)porphyrin ≥96.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
5,10,15,20-Tetra(4-pyridyl)-21H,23H-porphine, meso-Tetra(4-pyridyl)porphine
CAS Number: 16834-13-2
Molecular Formula: C40H26N8
Molecular Weight: 618.70
Purity/Analysis Method: >96.0% (N)
Form: Crystal
Color: Purple
Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Bioscience, Porphyrins, Analytical Science, Porphines (Chelating Agents)
Other Product Names / Synonyms: 5,10,15,20-Tetra(4-pyridyl)-21H,23H-porphine
Formula:
C₄₀H₂₆N₈ MW: 618.69 g/mol |
MDL Number:
MFCD00009626 CAS Number: 16834-13-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...