o-Tolyl-3,5-xylyl ether ≥97.0%

Supplier: TCI America

T2213-25G T2213-5G
TCT2213-25GEA 819.86 CAD
TCT2213-25G TCT2213-5G
o-Tolyl-3,5-xylyl ether ≥97.0%
o-Tolyl-3,5-xylyl ether

CAS Number: 196604-20-3
Molecular Formula: C15H16O
Molecular Weight: 212.29
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 2,3',5'-Trimethyldiphenyl Ether

Formula: C₁₅H₁₆O
Density: 1.03 (20 °C)
CAS Number: 196604-20-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbdba88ee6e570cd74bc3279f5b492ed223849d8&vid=dbdba88ee6e570cd74bc3279f5b492ed223849d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR