1,2,3,4-Tetra-O-acetyl-ɑ-L-fucopyranose ≥98.0%
Supplier: TCI America
Synonyms:
1,2,3,4-Tetra-O-acetyl-α-L(−)-fucose, Tetra-O-acetyl-α-L(−)fucopyranose, ɑ-L-Fucopyranose 1,2,3,4-Tetraacetate
CAS Number: 64913-16-2
MDL Number: MFCD00069791
Molecular Formula: C14H20O9
Molecular Weight: 332.31
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 94
Specific rotation [a]20/D: -120 deg (C=1, CHCl3)
Storage Temperature: <0°C
Product Categories: Bioscience, Fucose, O-Substituted Sugars
Other Product Names / Synonyms: alpha-L-Fucopyranose 1,2,3,4-Tetraacetate
Formula:
C₁₄H₂₀O₉ MW: 332.31 g/mol Melting Pt: 94 °C |
MDL Number:
MFCD00069791 CAS Number: 64913-16-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...