1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-β-D-glucopyranose ≥97.0% (by HPLC)

Supplier: TCI America

T2196-1G T2196-200MG
TCT2196-1GEA 629.79 CAD
TCT2196-1G TCT2196-200MG
1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-β-D-glucopyranose ≥97.0% (by HPLC)
1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-beta-D-glucopyranose

CAS Number: 80321-89-7
Molecular Formula: C14H19N3O9
Molecular Weight: 373.32
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 97
Specific rotation [a]20/D: 3 deg (C=1, CHCl3)
Storage Temperature: <0°C

Formula: C₁₄H₁₉N₃O₉
Melting Pt: 97 °C
CAS Number: 80321-89-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1e4acaf14e22ea03576919e63316ecab779c392&vid=a1e4acaf14e22ea03576919e63316ecab779c392&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR