Thiomorpholine-1,1-dioxide ≥98.0%
Supplier: TCI America
Synonyms:
Thiomorpholine 1,1-dioxide
T2193-5G
T2193-25G
TCT2193-5GEA
280.92
CAD
TCT2193-5G
TCT2193-25G
Thiomorpholine-1,1-dioxide ≥98.0%
Thiomorpholine 1,1-dioxide
CAS Number: 39093-93-1
Molecular Formula: C4H9NO2S
Molecular Weight: 135.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Boiling point (°C): 155
Melting point (°C): 70
Other Product Names / Synonyms: 1,1-Dioxothiomorpholine
Formula:
C₄H₉NO₂S MW: 135.1848 g/mol Boiling Pt: 155 °C Melting Pt: 70 °C |
MDL Number:
MFCD05861623 CAS Number: 39093-93-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...