2,4,5-Trifluoro-1,3-phenylenediamine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
T2060-5G
T2060-1G
TCT2060-5GEA
371.3
CAD
TCT2060-5G
TCT2060-1G
2,4,5-Trifluoro-1,3-phenylenediamine ≥98.0% (by titrimetric analysis)
2,4,5-Trifluoro-1,3-phenylenediamine
CAS Number: 321182-37-0
MDL Number: MFCD04114298
Molecular Formula: C6H5F3N2
Molecular Weight: 162.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1,3-Diamino-2,4,5-trifluorobenzene
Formula:
C₆H₅F₃N₂ |
MDL Number:
MFCD04114298 CAS Number: 321182-37-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...