2,4,6-Tris[bis(methoxymethyl)amino]-1,3,5-triazine ≥98.0%

Supplier: TCI America

T2059-25G T2059-5G
TCT2059-25GEA 500.86 CAD
TCT2059-25G TCT2059-5G
2,4,6-Tris[bis(methoxymethyl)amino]-1,3,5-triazine ≥98.0%
2,4,6-Tris[bis(methoxymethyl)amino]-1,3,5-triazine

CAS Number: 3089-11-0
MDL Number: MFCD00047342
Molecular Formula: C15H30N6O6
Molecular Weight: 390.44
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 52
Flash Point (°C): 197


Other Product Names / Synonyms: N,N,N',N',N'',N''-Hexakis(methoxymethyl)melamine

Formula: C₁₅H₃₀N₆O₆
Melting Pt: 52 °C
Flash Pt: 197 °C
MDL Number: MFCD00047342
CAS Number: 3089-11-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3adba4fec21ae5f2410d37083f2f04d10a8f7923&vid=3adba4fec21ae5f2410d37083f2f04d10a8f7923&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR