1,3,4,6-Tetrakis(methoxymethyl)glycoluril ≥98.0%

Supplier: TCI America

Synonyms: 1,3,4,6-Tetrakis(methoxymethyl)tetrahydroimidazo[4,5-d]imidazole-2,5(1H,3H)-dione

T2058-25G T2058-5G
TCT2058-25GEA 339.7 CAD
TCT2058-25G TCT2058-5G
1,3,4,6-Tetrakis(methoxymethyl)glycoluril ≥98.0%
1,3,4,6-Tetrakis(methoxymethyl)glycoluril

CAS Number: 17464-88-9
MDL Number: MFCD00402438
Molecular Formula: C12H22N4O6
Molecular Weight: 318.33
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Color: White
Melting point (°C): 118

Formula: C₁₂H₂₂N₄O₆
MW: 318.33 g/mol
Melting Pt: 118 °C
MDL Number: MFCD00402438
CAS Number: 17464-88-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f88e5b4ef5566efc2cc867e2ee21a31fad09acc2&vid=f88e5b4ef5566efc2cc867e2ee21a31fad09acc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR