2,3,4,6-Tetra-O-benzoyl-D-mannopyranose ≥93.0% (by HPLC)
Supplier: TCI America
T2056-1G
T2056-5G
TCT2056-1GEA
239.69
CAD
TCT2056-1G
TCT2056-5G
2,3,4,6-Tetra-O-benzoyl-D-mannopyranose ≥93.0% (by HPLC)
2,3,4,6-Tetra-O-benzoyl-D-mannopyranose
CAS Number: 113544-59-5
MDL Number: MFCD02683399
Molecular Formula: C34H28O10
Molecular Weight: 596.59
Purity/Analysis Method: >93.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 182
Specific rotation [a]20/D: -78 deg (C=1, CHCl3)
Storage Temperature: <0°C
Formula:
C₃₄H₂₈O₁₀ Melting Pt: 182 °C |
MDL Number:
MFCD02683399 CAS Number: 113544-59-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...