1,1,1',1'-Tetramethyl-4,4'-(methylenedi-p-phenylene)disemicarbazide ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

T2051-500G T2051-25G
TCT2051-500GEA 489.8 CAD
TCT2051-500G TCT2051-25G
1,1,1',1'-Tetramethyl-4,4'-(methylenedi-p-phenylene)disemicarbazide ≥95.0% (by HPLC, titration analysis)
1,1,1',1'-Tetramethyl-4,4'-(methylenedi-p-phenylene)disemicarbazide

CAS Number: 85095-61-0
MDL Number: MFCD00525631
Molecular Formula: C19H26N6O2
Molecular Weight: 370.46
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 177


Other Product Names / Synonyms: Bis[4-[3-(dimethylamino)ureido]phenyl]methane

Formula: C₁₉H₂₆N₆O₂
Melting Pt: 177 °C
MDL Number: MFCD00525631
CAS Number: 85095-61-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=904dc1bff316d9cb6e2f73562283fe62f95a6178&vid=904dc1bff316d9cb6e2f73562283fe62f95a6178&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR