Thiomorpholinoacetic acid-1',1'-dioxide monohydrate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
T2050-5G
T2050-25G
TCT2050-5GEA
167.48
CAD
TCT2050-5G
TCT2050-25G
Thiomorpholinoacetic acid-1',1'-dioxide monohydrate ≥98.0% (by GC, titration analysis)
Thiomorpholinoacetic acid-1',1'-dioxide monohydrate
CAS Number: 155480-08-3
MDL Number: MFCD00121261
Molecular Formula: C6H11NO4S
Molecular Weight: 193.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Color: White
Melting point (°C): 177
Other Product Names / Synonyms: 4-(Carboxymethyl)thiomorpholine 1,1-Dioxide, (1,1-Dioxothiomorpholino)acetic Acid
Formula:
C₆H₁₁NO₄S Melting Pt: 177 °C |
MDL Number:
MFCD00121261 CAS Number: 155480-08-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...