2-(2-Thienyl)-1,3-dioxolane ≥97.0%

Supplier: TCI America

T2049-5G T2049-25G
TCT2049-5GEA 94.8 CAD
TCT2049-5G TCT2049-25G
2-(2-Thienyl)-1,3-dioxolane ≥97.0%
2-(2-Thienyl)-1,3-dioxolane

CAS Number: 58268-08-9
MDL Number: MFCD01924576
Molecular Formula: C7H8O2S
Molecular Weight: 156.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 111
Specific Gravity (20/20): 1.25
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-(1,3-Dioxolan-2-yl)thiophene

Formula: C₇H₈O₂S
Boiling Pt: 111 °C
Density: 1.25 (20 °C)
MDL Number: MFCD01924576
CAS Number: 58268-08-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=06676dccff5e9c56b386dd106c1d7b110623b5ae&vid=06676dccff5e9c56b386dd106c1d7b110623b5ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR