N-Chloromethyl-4-nitrophthalimide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

A5503-5G A5503-1G
TCA5503-5GEA 750.5 CAD
TCA5503-5G TCA5503-001G
N-Chloromethyl-4-nitrophthalimide ≥98.0% (by titrimetric analysis)
N-Chloromethyl-4-nitrophthalimide

[for HPLC Labeling]
CAS Number: 54455-34-4
MDL Number: MFCD00059889
Molecular Formula: C9H5ClN2O4
Molecular Weight: 240.60
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Analytical Science, UV Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 4-Nitro-N-chloromethylphthalimide

Formula: C₉H₅ClN₂O₄
MDL Number: MFCD00059889
CAS Number: 54455-34-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02f5ce18560bdc1a7cd1b54e24ff4980d34ae87c&vid=02f5ce18560bdc1a7cd1b54e24ff4980d34ae87c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR