1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-β-D-glucopyranose ≥97.0% (by HPLC)

Supplier: TCI America

T2047-25G T2047-5G
TCT2047-25GEA 368.61 CAD
TCT2047-25G TCT2047-5G
1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-β-D-glucopyranose ≥97.0% (by HPLC)
1,3,4,6-Tetra-O-acetyl-2-deoxy-2-phthalimido-beta-D-glucopyranose

CAS Number: 10022-13-6
Molecular Formula: C22H23NO11
Molecular Weight: 477.42
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 200
Specific rotation [a]20/D: 68 deg (C=1, CHCl3)
Storage Temperature: <0°C

Formula: C₂₂H₂₃NO₁₁
Melting Pt: 200 °C
MDL Number: MFCD00080781
CAS Number: 10022-13-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6c778ecab87175abc2e0ccd476033e0b08a03e3&vid=d6c778ecab87175abc2e0ccd476033e0b08a03e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR