Tri-p-tolylsulfonium trifluoromethanesulfonate ≥95.0%
Supplier: TCI America
Synonyms:
Tris(4-methylphenyl)sulfonium Trifluoromethanesulfonate, Tris(4-methylphenyl)sulphonium Trifluoromethanesulphonate, Tri-p-tolylsulfonium Triflate, Tri-p-tolylsulphonium Triflate
CAS Number: 127820-38-6
MDL Number: MFCD06797188
Molecular Formula: C22H21F3O3S2
Molecular Weight: 454.52
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 106
Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: Tris(4-methylphenyl)sulfonium Trifluoromethanesulfonate, Tri-p-tolylsulfonium Triflate
Formula:
C₂₂H₂₁F₃O₃S₂ MW: 454.52 g/mol Melting Pt: 106 °C |
MDL Number:
MFCD06797188 CAS Number: 127820-38-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...