Sodium trifluoromethanesulfinate ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Trifluoromethanesulphinic acid Sodium Salt, Langlois Reagent, Trifluoromethanesulfinic acid Sodium Salt

T2033-25G T2033-5G
TCT2033-25GEA 399.74 CAD
TCT2033-25G TCT2033-5G
Sodium trifluoromethanesulfinate ≥95.0% (by titrimetric analysis)
Sodium trifluoromethanesulfinate

CAS Number: 2926-29-6
MDL Number: MFCD03092989
Molecular Formula: CHF3O2S
Molecular Weight: 156.05
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Color: White


Product Categories: Synthetic Organic Chemistry, Fluorinating Agents, Building Blocks, Fluorinated Bioactive Compounds
Other Product Names / Synonyms: Trifluoromethanesulfinic Acid Sodium Salt, Langlois Reagent

Formula: CHF₃O₂S
MW: 156.05 g/mol
MDL Number: MFCD03092989
CAS Number: 2926-29-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c8ef3e989609a2d4f36a1327d5ea716631257a8&vid=5c8ef3e989609a2d4f36a1327d5ea716631257a8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR