2,3,4,6-Tetra-O-benzoyl-D-glucopyranose ≥90.0%

Supplier: TCI America

Synonyms: 2,3,4,6-Tetrabenzoyl-D-glucopyranose

T2020-1G T2020-250MG
TCT2020-1GEA 693.62 CAD
TCT2020-1G TCT2020-250MG
2,3,4,6-Tetra-O-benzoyl-D-glucopyranose ≥90.0%
2,3,4,6-Tetra-O-benzoyl-D-glucopyranose

CAS Number: 64768-20-3
MDL Number: MFCD00066004
Molecular Formula: C34H28O10
Molecular Weight: 596.59
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal
Color: White
Melting point (°C): 122
Specific rotation [a]20/D: 73 deg (C=1, Dioxane)

Formula: C₃₄H₂₈O₁₀
MW: 596.59 g/mol
Melting Pt: 122 °C
MDL Number: MFCD00066004
CAS Number: 64768-20-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d16605886dbe97008f6d5e04d247223cd379fef8&vid=d16605886dbe97008f6d5e04d247223cd379fef8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR