PTIO (=2-phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl) ≥98.0%

Supplier: TCI America

Synonyms: 2-Phenyl-4,4,5,5-tetramethylimidazoline-1-oxyl 3-oxide, 4,5-Dihydro-4,4,5,5-tetramethyl-2-phenyl-1H-imidazol-1-yloxy-1-oxide, 2-Phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl, PTIO

A5440-1G A5440-5G
TCA5440-001GEA 356.27 CAD
TCA5440-001G TCA5440-005G
PTIO (=2-phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl) ≥98.0%
PTIO (=2-phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl)

[Stable free radical reagent for the simultaneous determination of NO and NO2 in the atmosphere]
CAS Number: 18390-00-6
MDL Number: MFCD00059929
Molecular Formula: C13H17N2O2
Molecular Weight: 233.29
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Bioscience, ROS/RNS Detection Reagents, Reagents for Nitric Oxide Research, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: 4,5-Dihydro-4,4,5,5-tetramethyl-2-phenyl-1H-imidazol-1-yloxy-1-oxide, 2-Phenyl-4,4,5,5-tetramethylimidazoline-3-oxide-1-oxyl

Formula: C₁₃H₁₇N₂O₂
MW: 233.29 g/mol
MDL Number: MFCD00059929
CAS Number: 18390-00-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a7235dc6217810c5f26553fd3a493eb6990e380&vid=5a7235dc6217810c5f26553fd3a493eb6990e380&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR