2,4,5,6-Tetrafluoro-1,3-phenylenediamine ≥95.0%
Supplier: TCI America
Synonyms:
1,3-Diamino-2,4,5,6-tetrafluorobenzene
T2019-1G
T2019-5G
TCT2019-1GEA
199.08
CAD
TCT2019-1G
TCT2019-5G
2,4,5,6-Tetrafluoro-1,3-phenylenediamine ≥95.0%
2,4,5,6-Tetrafluoro-1,3-phenylenediamine
CAS Number: 1198-63-6
MDL Number: MFCD00039764
Molecular Formula: C6H4F4N2
Molecular Weight: 180.11
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Color: White
Melting point (°C): 131
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Materials Chemistry, Monomers, Polyimide Raw Materials
Other Product Names / Synonyms: 1,3-Diamino-2,4,5,6-tetrafluorobenzene
Formula:
C₆H₄F₄N₂ MW: 180.11 g/mol Melting Pt: 131 °C |
MDL Number:
MFCD00039764 CAS Number: 1198-63-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...