Tetrabutylphosphonium hexafluorophosphate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Tetrabutylphosphonium hexafluorophosphate(V)

T2007-25G T2007-5G
TCT2007-25GEA 330.54 CAD
TCT2007-25G TCT2007-5G
Tetrabutylphosphonium hexafluorophosphate ≥98.0% (by titrimetric analysis)
Tetrabutylphosphonium hexafluorophosphate

CAS Number: 111928-21-3
MDL Number: MFCD01863114
Molecular Formula: C16H36F6P2
Molecular Weight: 404.40
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Color: White
Melting point (°C): 217

Formula: C₁₆H₃₆F₆P₂
MW: 404.3950 g/mol
Melting Pt: 217 °C
MDL Number: MFCD01863114
CAS Number: 111928-21-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65109cb78eaf8e34e80f34a358f490e84ebd2bde&vid=65109cb78eaf8e34e80f34a358f490e84ebd2bde&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR