2,3,4,6-Tetra-O-benzyl-ɑ-D-glucopyranosyl-p-trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate

Supplier: TCI America

T1999-1G T1999-200MG
TCT1999-1GEA 628.05 CAD
TCT1999-1G TCT1999-200MG
2,3,4,6-Tetra-O-benzyl-ɑ-D-glucopyranosyl-p-trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate
2,3,4,6-Tetra-O-benzyl-alpha-D-glucopyranosyl-p-trifluoromethylbenzylthio-N-(p-trifluoromethylphenyl)formimidate

CAS Number: 468095-63-8
MDL Number: MFCD06797177
Molecular Formula: C50H45F6NO6S
Molecular Weight: 901.96
Form: Crystal
Color: White
Melting point (°C): 123
Specific rotation [a]20/D: 63 deg (C=1, CHCl3)
Storage Temperature: <0°C

Formula: C₅₀H₄₅F₆NO₆S
MW: 901.96 g/mol
Melting Pt: 123 °C
MDL Number: MFCD06797177
CAS Number: 468095-63-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d791eda250f572879c87e9e821004c2be948babe&vid=d791eda250f572879c87e9e821004c2be948babe&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR